missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Methyl Sulfone 98.0+%, TCI America™
Supplier: TCI America E0474100G
Specifications
Ethyl Methyl Sulfone | |
White | |
MFCD00015543 | |
Methylsulfonylethane | |
CCS(C)(=O)=O | |
108.16 | |
108.16 | |
Crystals |
594-43-4 | |
C3H8O2S | |
100 g | |
YBJCDTIWNDBNTM-UHFFFAOYSA-N | |
methanesulfonylethane | |
79059 | |
≥98.0% (GC) |
Chemical Identifiers
594-43-4 | |
108.16 | |
YBJCDTIWNDBNTM-UHFFFAOYSA-N | |
79059 | |
CCS(C)(=O)=O |
C3H8O2S | |
MFCD00015543 | |
Methylsulfonylethane | |
methanesulfonylethane |
Safety and Handling
TSCA : No