missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl N-phenylformimidate, 97%, Thermo Scientific Chemicals
$51.14 - $161.99
Chemical Identifiers
CAS | 6780-49-0 |
---|---|
Molecular Formula | C9H11NO |
Molecular Weight (g/mol) | 149.19 |
MDL Number | MFCD00043655 |
InChI Key | DRDBNKYFCOLNQO-UHFFFAOYSA-N |
Synonym | ethyl n-phenylformimidate, ethyl isoformanilide, n-ethoxymethyleneaniline, methanimidic acid, n-phenyl-, ethyl ester, ethyl phenylimidoformate, n-ethoxymethylene aniline, ethyl n-phenylcarboximidate, ethylisoformanilide, e-ethyl n-phenylcarboximidate, ethyl iso-formanilide |
PubChem CID | 111236 |
IUPAC Name | ethyl N-phenylmethanimidate |
SMILES | CCOC=NC1=CC=CC=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6780-49-0 | |
149.19 | |
DRDBNKYFCOLNQO-UHFFFAOYSA-N | |
111236 | |
CCOC=NC1=CC=CC=C1 |
C9H11NO | |
MFCD00043655 | |
ethyl n-phenylformimidate, ethyl isoformanilide, n-ethoxymethyleneaniline, methanimidic acid, n-phenyl-, ethyl ester, ethyl phenylimidoformate, n-ethoxymethylene aniline, ethyl n-phenylcarboximidate, ethylisoformanilide, e-ethyl n-phenylcarboximidate, ethyl iso-formanilide | |
ethyl N-phenylmethanimidate |
Specifications
6780-49-0 | |
101°C to 102°C (15 mmHg) | |
MFCD00043655 | |
Air and moisture sensitive | |
DRDBNKYFCOLNQO-UHFFFAOYSA-N | |
ethyl N-phenylmethanimidate | |
111236 | |
97% | |
Ethyl N-phenylformimidate |
Colorless | |
C9H11NO | |
5 g | |
ethyl n-phenylformimidate, ethyl isoformanilide, n-ethoxymethyleneaniline, methanimidic acid, n-phenyl-, ethyl ester, ethyl phenylimidoformate, n-ethoxymethylene aniline, ethyl n-phenylcarboximidate, ethylisoformanilide, e-ethyl n-phenylcarboximidate, ethyl iso-formanilide | |
CCOC=NC1=CC=CC=C1 | |
149.19 | |
149.19 | |
Liquid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 229-841-3
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only