Learn More
Ethyl N-piperazinecarboxylate, 99%
CAS: 120-43-4 | C7H14N2O2 | 158.2 g/mol
$62.08 - $222.22
Chemical Identifiers
| CAS | 120-43-4 |
|---|---|
| Molecular Formula | C7H14N2O2 |
| Molecular Weight (g/mol) | 158.2 |
| MDL Number | MFCD00005964 |
| InChI Key | LNOQURRKNJKKBU-UHFFFAOYSA-N |
| Synonym | ethyl n-piperazinecarboxylate, ethyl 1-piperazinecarboxylate, n-carbethoxypiperazine, 1-carbethoxypiperazine, 1-carboethoxypiperazine, 1-piperazinecarboxylic acid, ethyl ester, n-ethoxycarbonyl piperazine, ethylcarbonyl piperazine, 1-ethoxycarbonyl piperazine, ethyl 1-piperazinocarboxylate |
| PubChem CID | 8431 |
| IUPAC Name | ethyl piperazine-1-carboxylate |
| SMILES | CCOC(=O)N1CCNCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC118870250
|
Thermo Scientific Chemicals
118870250 |
25 g |
Each for $62.08
|
|
|||||
|
AC118871000
|
Thermo Scientific Chemicals
118871000 |
100 g |
Each for $222.22
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 120-43-4 | |
| 158.2 | |
| LNOQURRKNJKKBU-UHFFFAOYSA-N | |
| 8431 | |
| CCOC(=O)N1CCNCC1 |
| C7H14N2O2 | |
| MFCD00005964 | |
| ethyl n-piperazinecarboxylate, ethyl 1-piperazinecarboxylate, n-carbethoxypiperazine, 1-carbethoxypiperazine, 1-carboethoxypiperazine, 1-piperazinecarboxylic acid, ethyl ester, n-ethoxycarbonyl piperazine, ethylcarbonyl piperazine, 1-ethoxycarbonyl piperazine, ethyl 1-piperazinocarboxylate | |
| ethyl piperazine-1-carboxylate |
Specifications
| 120-43-4 | |
| 1.0800g/mL | |
| >110°C | |
| 98.5% min. (GC) | |
| C7H14N2O2 | |
| MFCD00005964 | |
| 23, II, 9 | |
| ethyl n-piperazinecarboxylate, ethyl 1-piperazinecarboxylate, n-carbethoxypiperazine, 1-carbethoxypiperazine, 1-carboethoxypiperazine, 1-piperazinecarboxylic acid, ethyl ester, n-ethoxycarbonyl piperazine, ethylcarbonyl piperazine, 1-ethoxycarbonyl piperazine, ethyl 1-piperazinocarboxylate | |
| LNOQURRKNJKKBU-UHFFFAOYSA-N | |
| ethyl piperazine-1-carboxylate | |
| 8431 | |
| 99% | |
| Ethyl N-piperazinecarboxylate |
| Yellow | |
| 273°C | |
| Authentic | |
| Glass Bottle | |
| 1.4760 to 1.478 | |
| 25 g | |
| 1.08 | |
| Solubility in water: freely soluble | |
| CCOC(=O)N1CCNCC1 | |
| 158.2 | |
| 158.2 | |
| Oily Liquid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 204-395-2
RUO – Research Use Only