Learn More
Ethyl nicotinate, 99%, Thermo Scientific Chemicals
$79.81 - $220.97
Chemical Identifiers
CAS | 614-18-6 |
---|---|
Molecular Formula | C8H9NO2 |
Molecular Weight (g/mol) | 151.165 |
MDL Number | MFCD00006389 |
InChI Key | XBLVHTDFJBKJLG-UHFFFAOYSA-N |
Synonym | ethyl nicotinate, ethyl nicotinoate, nicotinic acid ethyl ester, ethyl 3-pyridinecarboxylate, 3-pyridinecarboxylic acid, ethyl ester, ethylnicotinate, nicotinic acid, ethyl ester, mucotherm, nicaethan, nikethan |
PubChem CID | 69188 |
IUPAC Name | ethyl pyridine-3-carboxylate |
SMILES | CCOC(=O)C1=CN=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0321522
|
Thermo Scientific Chemicals
L0321522 |
100 g |
Each for $79.81
|
|
||||||
AAL0321536
|
Thermo Scientific Chemicals
L0321536 |
500 g |
Each for $220.97
|
|
||||||
Description
Ethyl nicotinate is used for skin-conditioning cosmetics. It is principally used in medicine as a local vasodilator at a concentration of 1 to 2 % in creams, ointments and pomades. It is utilized for the treatment sprains, wrenches, muscular pains and tendonitis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl nicotinate is used for skin-conditioning cosmetics. It is principally used in medicine as a local vasodilator at a concentration of 1 to 2 % in creams, ointments and pomades. It is utilized for the treatment sprains, wrenches, muscular pains and tendonitis.
Solubility
Miscible with water.
Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
614-18-6 | |
151.165 | |
XBLVHTDFJBKJLG-UHFFFAOYSA-N | |
69188 | |
CCOC(=O)C1=CN=CC=C1 |
C8H9NO2 | |
MFCD00006389 | |
ethyl nicotinate, ethyl nicotinoate, nicotinic acid ethyl ester, ethyl 3-pyridinecarboxylate, 3-pyridinecarboxylic acid, ethyl ester, ethylnicotinate, nicotinic acid, ethyl ester, mucotherm, nicaethan, nikethan | |
ethyl pyridine-3-carboxylate |
Specifications
614-18-6 | |
1.107 | |
93°C (199°F) | |
C8H9NO2 | |
MFCD00006389 | |
122937 | |
Miscible with water. | |
CCOC(=O)C1=CN=CC=C1 | |
151.165 | |
151.17 | |
Ethyl nicotinate |
8°C to 9°C | |
223°C to 224°C | |
Characteristic | |
1.503 | |
100 g | |
ethyl nicotinate, ethyl nicotinoate, nicotinic acid ethyl ester, ethyl 3-pyridinecarboxylate, 3-pyridinecarboxylic acid, ethyl ester, ethylnicotinate, nicotinic acid, ethyl ester, mucotherm, nicaethan, nikethan | |
XBLVHTDFJBKJLG-UHFFFAOYSA-N | |
ethyl pyridine-3-carboxylate | |
69188 | |
99% |
Safety and Handling
GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H500
EINECSNumber : 210-370-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only