missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl oxamate, 99%, Thermo Scientific Chemicals
$38.18 - $38.18
Chemical Identifiers
CAS | 617-36-7 |
---|---|
MDL Number | MFCD00008005 |
InChI Key | RZMZBHSKPLVQCP-UHFFFAOYSA-N |
Synonym | ethyl oxamate, ethyl carbamoylformate, oxamic acid ethyl ester, ethyl amino oxo acetate, oxamethane, acetic acid, aminooxo-, ethyl ester, ethoxalamide, oxamidic acid ethyl ester, ethoxy carbonyl formamide, oxalic acid, monoethyl ester amide |
PubChem CID | 69238 |
IUPAC Name | ethyl 2-amino-2-oxoacetate |
SMILES | CCOC(=O)C(=O)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC118790250
|
Thermo Scientific Chemicals
118790250 |
25 g | Glass bottle |
Each for $38.18
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
617-36-7 | |
RZMZBHSKPLVQCP-UHFFFAOYSA-N | |
69238 | |
CCOC(=O)C(=O)N |
MFCD00008005 | |
ethyl oxamate, ethyl carbamoylformate, oxamic acid ethyl ester, ethyl amino oxo acetate, oxamethane, acetic acid, aminooxo-, ethyl ester, ethoxalamide, oxamidic acid ethyl ester, ethoxy carbonyl formamide, oxalic acid, monoethyl ester amide | |
ethyl 2-amino-2-oxoacetate |
Specifications
617-36-7 | |
White | |
98.5% min | |
C4H7NO3 | |
MFCD00008005 | |
02, 544 | |
Solubility in water: soluble | |
CCOC(=O)C(=O)N | |
117.1 | |
117.1 | |
Crystalline Powder, Crystals or Chunks |
112°C to 115°C | |
Authentic | |
Glass bottle | |
H2NCOCO2C2H5 | |
25 g | |
ethyl oxamate, ethyl carbamoylformate, oxamic acid ethyl ester, ethyl amino oxo acetate, oxamethane, acetic acid, aminooxo-, ethyl ester, ethoxalamide, oxamidic acid ethyl ester, ethoxy carbonyl formamide, oxalic acid, monoethyl ester amide | |
RZMZBHSKPLVQCP-UHFFFAOYSA-N | |
ethyl 2-amino-2-oxoacetate | |
69238 | |
99% | |
Ethyl oxamate |
Safety and Handling
EINECSNumber : 210-512-8
RUO – Research Use Only