missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl oxazole-5-carboxylate, 98%
CAS: 118994-89-1 | C6H7NO3 | 141.126 g/mol
$173.16 - $541.95
Chemical Identifiers
| CAS | 118994-89-1 |
|---|---|
| Molecular Formula | C6H7NO3 |
| Molecular Weight (g/mol) | 141.126 |
| MDL Number | MFCD04114930 |
| InChI Key | KRMORCCAHXFIHF-UHFFFAOYSA-N |
| Synonym | ethyl oxazole-5-carboxylate, 5-oxazolecarboxylic acid, ethyl ester, 5-oxazolecarboxylicacid, ethyl ester, oxazole-5-carboxylic acid ethyl ester, ethyloxazole-5-carboxylate, 5-ethoxycarbonyloxazole, acmc-1c1iz, ksc510s9l, ethyl 5-oxazolecarboxylate, 5-ethoxycarbonyl-1,3-oxazole |
| PubChem CID | 10964603 |
| IUPAC Name | ethyl 1,3-oxazole-5-carboxylate |
| SMILES | CCOC(=O)C1=CN=CO1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3292406
|
Thermo Scientific Chemicals
H3292406 |
5 g |
N/A
|
|
|||||
|
AAH3292414
|
Thermo Scientific Chemicals
H3292414 |
25 g |
N/A
|
|
|||||
Description
Ethyl oxazole-5-carboxylate is used as a pharmaceutical, organic intermediate and in organic light-emitting diode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl oxazole-5-carboxylate is used as a pharmaceutical, organic intermediate and in organic light-emitting diode.
Solubility
Soluble in water (35 g/L at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| 118994-89-1 | |
| 141.126 | |
| KRMORCCAHXFIHF-UHFFFAOYSA-N | |
| 10964603 | |
| CCOC(=O)C1=CN=CO1 |
| C6H7NO3 | |
| MFCD04114930 | |
| ethyl oxazole-5-carboxylate, 5-oxazolecarboxylic acid, ethyl ester, 5-oxazolecarboxylicacid, ethyl ester, oxazole-5-carboxylic acid ethyl ester, ethyloxazole-5-carboxylate, 5-ethoxycarbonyloxazole, acmc-1c1iz, ksc510s9l, ethyl 5-oxazolecarboxylate, 5-ethoxycarbonyl-1,3-oxazole | |
| ethyl 1,3-oxazole-5-carboxylate |
Specifications
| 118994-89-1 | |
| C6H7NO3 | |
| MFCD04114930 | |
| 83084416 | |
| Soluble in water (35g/L at 25°C). | |
| CCOC(=O)C1=CN=CO1 | |
| 141.126 | |
| 141.13 | |
| Ethyl oxazole-5-carboxylate |
| 60°C (2.25 mmHg) | |
| 1.469 | |
| 5 g | |
| ethyl oxazole-5-carboxylate, 5-oxazolecarboxylic acid, ethyl ester, 5-oxazolecarboxylicacid, ethyl ester, oxazole-5-carboxylic acid ethyl ester, ethyloxazole-5-carboxylate, 5-ethoxycarbonyloxazole, acmc-1c1iz, ksc510s9l, ethyl 5-oxazolecarboxylate, 5-ethoxycarbonyl-1,3-oxazole | |
| KRMORCCAHXFIHF-UHFFFAOYSA-N | |
| ethyl 1,3-oxazole-5-carboxylate | |
| 10964603 | |
| 98% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only