missing translation for 'onlineSavingsMsg'
Learn More

Ethyl p-toluenesulfonate, 98%

CAS: 80-40-0 | C9H12O3S | 200.25 g/mol

$53.29 - $53.29

Chemical Identifiers

CAS 80-40-0
Molecular Formula C9H12O3S
Molecular Weight (g/mol) 200.25
MDL Number MFCD00009100
InChI Key VRZVPALEJCLXPR-UHFFFAOYSA-N
Synonym ethyl p-toluenesulfonate, ethyl tosylate, ethyl p-tosylate, ethyl p-ts, benzenesulfonic acid, 4-methyl-, ethyl ester, ethyl p-methylbenzenesulfonate, ethyl-p-toluenesulfonate, p-toluenesulfonic acid, ethyl ester, ethyl pts, p-toluolsulfonsaeure aethyl ester
PubChem CID 6638
IUPAC Name ethyl 4-methylbenzenesulfonate
SMILES CCOS(=O)(=O)C1=CC=C(C=C1)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC147230500
SDS View Documents
Thermo Scientific Chemicals
147230500
50 g Glass bottle
Each for $53.29
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

80-40-0
200.25
VRZVPALEJCLXPR-UHFFFAOYSA-N
6638
CCOS(=O)(=O)C1=CC=C(C=C1)C
C9H12O3S
MFCD00009100
ethyl p-toluenesulfonate, ethyl tosylate, ethyl p-tosylate, ethyl p-ts, benzenesulfonic acid, 4-methyl-, ethyl ester, ethyl p-methylbenzenesulfonate, ethyl-p-toluenesulfonate, p-toluenesulfonic acid, ethyl ester, ethyl pts, p-toluolsulfonsaeure aethyl ester
ethyl 4-methylbenzenesulfonate

Specifications

80-40-0
Colorless to Brown
158.0°C to 162.0°C (10.0 mmHg)
Authentic
Glass bottle
1.5100 to 1.5120
MFCD00009100
11, 99
15, 3907
Solubility in water: insoluble. Other solubilities: soluble in many organic solvents
CCOS(=O)(=O)C1=CC=C(C=C1)C
200.25
200.25
Liquid After Melting
32.0°C to 34.0°C
1.1740g/mL
157°C
97.5% min. (GC)
C9H12O3S
CH3C6H4SO3C2H5
50 g
1.174
ethyl p-toluenesulfonate, ethyl tosylate, ethyl p-tosylate, ethyl p-ts, benzenesulfonic acid, 4-methyl-, ethyl ester, ethyl p-methylbenzenesulfonate, ethyl-p-toluenesulfonate, p-toluenesulfonic acid, ethyl ester, ethyl pts, p-toluolsulfonsaeure aethyl ester
VRZVPALEJCLXPR-UHFFFAOYSA-N
ethyl 4-methylbenzenesulfonate
6638
98%
Ethyl p-toluenesulfonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing genetic defects.
Suspected of causing cancer.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-276-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only