missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenylglyoxylate, 98%

CAS: 1603-79-8 | C10H10O3 | 178.19 g/mol

$59.14 - $510.18

Chemical Identifiers

CAS 1603-79-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00009120
InChI Key QKLCQKPAECHXCQ-UHFFFAOYSA-N
Synonym ethyl benzoylformate, ethyl phenylglyoxylate, ethyl oxophenylacetate, ethylbenzoylformate, phenylglyoxylic acid ethyl ester, ethyl oxo phenyl acetate, benzeneacetic acid, .alpha.-oxo-, ethyl ester, benzoylformic acid ethyl ester, ethyl benzoyl formate, glyoxylic acid, phenyl-, ethyl ester
PubChem CID 15349
ChEBI CHEBI:84260
IUPAC Name ethyl 2-oxo-2-phenylacetate
SMILES CCOC(=O)C(=O)C1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2529206
View Documents
Thermo Scientific Chemicals
B2529206
5 g
Each for $59.14
Only null left
 
AAB2529214
View Documents
Thermo Scientific Chemicals
B2529214
25 g
Each for $151.29
Only null left
 
AAB2529222
View Documents
Thermo Scientific Chemicals
B2529222
100 g
Each for $510.18
Only null left
 
Description

Description

Ethyl phenylglyoxylate has been used in preparation of 1,5-dihydro-5-deazaflavin derivatives possessing a chiral substituent at N(3) position.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl phenylglyoxylate has been used in preparation of 1,5-dihydro-5-deazaflavin derivatives possessing a chiral substituent at N(3) position.

Solubility
Soluble in water 1143 mg/L @ 25°C .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications

Chemical Identifiers

1603-79-8
178.19
QKLCQKPAECHXCQ-UHFFFAOYSA-N
15349
ethyl 2-oxo-2-phenylacetate
C10H10O3
MFCD00009120
ethyl benzoylformate, ethyl phenylglyoxylate, ethyl oxophenylacetate, ethylbenzoylformate, phenylglyoxylic acid ethyl ester, ethyl oxo phenyl acetate, benzeneacetic acid, .alpha.-oxo-, ethyl ester, benzoylformic acid ethyl ester, ethyl benzoyl formate, glyoxylic acid, phenyl-, ethyl ester
CHEBI:84260
CCOC(=O)C(=O)C1=CC=CC=C1

Specifications

1603-79-8
138°C to 139°C (18 mmHg)
C10H10O3
MFCD00009120
Moisture sensitive
Soluble in water 1143mg/L @ 25°C .
CCOC(=O)C(=O)C1=CC=CC=C1
178.19
CHEBI:84260
98%
1.122
>110°C (230°F)
1.516
5 g
ethyl benzoylformate, ethyl phenylglyoxylate, ethyl oxophenylacetate, ethylbenzoylformate, phenylglyoxylic acid ethyl ester, ethyl oxo phenyl acetate, benzeneacetic acid, .alpha.-oxo-, ethyl ester, benzoylformic acid ethyl ester, ethyl benzoyl formate, glyoxylic acid, phenyl-, ethyl ester
QKLCQKPAECHXCQ-UHFFFAOYSA-N
ethyl 2-oxo-2-phenylacetate
15349
178.19
Ethyl phenylglyoxylate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-504-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only