missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl potassium malonate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 248080250

 View more versions of this product

Catalog No. AC248080250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl potassium malonate
6148-64-7
White
98%
C5H7KO4
MFCD00035603
02,III,1609
ethyl potassium malonate, potassium 3-ethoxy-3-oxopropanoate, potassium ethyl malonate, potassium monoethyl malonate, monoethyl malonate potassium salt, malonic acid monoethyl ester potassium salt, monoethyl potassium malonate, ethyl malonate potassium salt, propanedioic acid, monoethyl ester, potassium salt, ethylpotassiummalonate
WVUCPRGADMCTBN-UHFFFAOYSA-M
potassium;3-ethoxy-3-oxopropanoate
3446434
98%
98%
200.0°C
Authentic
Glass bottle
KOOCCH2COOC2H5
25 g
15,156
Solubility in water: 2.53g/L (25°C)
CCOC(=O)CC(=O)[O-].[K+]
170.2
170.2
Crystals or Powder

Chemical Identifiers

6148-64-7
170.2
WVUCPRGADMCTBN-UHFFFAOYSA-M
3446434
CCOC(=O)CC(=O)[O-].[K+]
C5H7KO4
MFCD00035603
ethyl potassium malonate, potassium 3-ethoxy-3-oxopropanoate, potassium ethyl malonate, potassium monoethyl malonate, monoethyl malonate potassium salt, malonic acid monoethyl ester potassium salt, monoethyl potassium malonate, ethyl malonate potassium salt, propanedioic acid, monoethyl ester, potassium salt, ethylpotassiummalonate
potassium;3-ethoxy-3-oxopropanoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-156-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only