missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl propenyl ether, 98%, mixture of cis- and trans-isomers, Thermo Scientific Chemicals

$104.85 - $104.85

Chemical Identifiers

CAS 928-55-2
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00015184
InChI Key XDHOEHJVXXTEDV-HYXAFXHYSA-N
Synonym 1-ethoxypropene, ethyl propenyl ether, ether, ethyl propenyl, propenyl ethyl ether, e-1-ethoxyprop-1-ene, 1-ethoxy-1-propene, unii-jh817due12, ethyl 1-propenyl ether, 1-propene, 1-ethoxy, ethyl-1-propenyl ether
PubChem CID 5365091
IUPAC Name (E)-1-ethoxyprop-1-ene
SMILES CCO\C=C/C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC410370100
SDS View Documents
Thermo Scientific Chemicals
410370100
10 g Glass bottle N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

928-55-2
86.13
XDHOEHJVXXTEDV-HYXAFXHYSA-N
5365091
CCO\C=C/C
C5H10O
MFCD00015184
1-ethoxypropene, ethyl propenyl ether, ether, ethyl propenyl, propenyl ethyl ether, e-1-ethoxyprop-1-ene, 1-ethoxy-1-propene, unii-jh817due12, ethyl 1-propenyl ether, 1-propene, 1-ethoxy, ethyl-1-propenyl ether
(E)-1-ethoxyprop-1-ene

Specifications

928-55-2
0.7780g/mL
−18°C
97.5% min. (GC)
C5H10O
CH3CHCHOCH2CH3
10 g
0.778
Solubility in water: soluble. Other solubilities: soluble in alkanes,methylene chloride,ethanol,,2-propanol,acetone
CCO\C=C/C
86.13
86.13
Liquid
Colorless to Yellow
67.0°C to 76.0°C
Authentic
Glass bottle
1.3940 to 1.4000
MFCD00015184
01,385
1-ethoxypropene, ethyl propenyl ether, ether, ethyl propenyl, propenyl ethyl ether, e-1-ethoxyprop-1-ene, 1-ethoxy-1-propene, unii-jh817due12, ethyl 1-propenyl ether, 1-propene, 1-ethoxy, ethyl-1-propenyl ether
XDHOEHJVXXTEDV-HYXAFXHYSA-N
(E)-1-ethoxyprop-1-ene
5365091
98%
Ethyl propenyl ether, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-176-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only