Learn More
Ethyl sorbate, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1143122
Description
The addition of chlorine and bromine to ethyl sorbate (la) gave 1, 2-and 1, a-dihalo products derived from attack of the halogen. Chlorination of la under ionic conditions proceeds through a tightly bridged chloronium ion intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe addition of chlorine and bromine to ethyl sorbate (la) gave 1, 2-and 1, a-dihalo products derived from attack of the halogen. Chlorination of la under ionic conditions proceeds through a tightly bridged chloronium ion intermediate.
Solubility
Soluble in alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Ethyl sorbate | |
0.948 | |
69°C (156°F) | |
1.495 | |
100 g | |
ethyl sorbate, ethyl 2,4-hexadienoate, ethyl hexa-2,4-dienoate, sorbic acid, ethyl ester, 2,4-hexadienoic acid, ethyl ester, ethyl 2e,4e-hexa-2,4-dienoate, ethyl e,e-2,4-hexadienoate, unii-hsr16usg4d, 2,4-hexadienoic acid, ethyl ester, 2e,4e, fema no. 2459 | |
OZZYKXXGCOLLLO-TWTPFVCWSA-N | |
ethyl (2E,4E)-hexa-2,4-dienoate | |
1550470 | |
98% |
2396-84-1 | |
195°C to 196°C | |
C8H12O2 | |
MFCD00009296 | |
1362941 | |
Soluble in alcohol. | |
CCOC(=O)\C=C\C=C\C | |
140.18 | |
140.18 |
Chemical Identifiers
2396-84-1 | |
140.18 | |
OZZYKXXGCOLLLO-TWTPFVCWSA-N | |
1550470 | |
CCOC(=O)\C=C\C=C\C |
C8H12O2 | |
MFCD00009296 | |
ethyl sorbate, ethyl 2,4-hexadienoate, ethyl hexa-2,4-dienoate, sorbic acid, ethyl ester, 2,4-hexadienoic acid, ethyl ester, ethyl 2e,4e-hexa-2,4-dienoate, ethyl e,e-2,4-hexadienoate, unii-hsr16usg4d, 2,4-hexadienoic acid, ethyl ester, 2e,4e, fema no. 2459 | |
ethyl (2E,4E)-hexa-2,4-dienoate |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 219-258-2
RTECSNumber : WG2143000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only