missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylaluminium dichloride, 0.9M solution in heptane, AcroSeal™, Thermo Scientific Chemicals

$166.13 - $166.13

Chemical Identifiers

CAS 563-43-9
Molecular Formula C2H5AlCl2
Molecular Weight (g/mol) 126.94
MDL Number MFCD00000457
InChI Key UAIZDWNSWGTKFZ-UHFFFAOYSA-L
Synonym ethylaluminum dichloride, aluminum, dichloroethyl, dichloroethylaluminum, ethyldichloroaluminum, dichloromonoethylaluminum, ethylaluminium dichloride, dichloro ethyl alumane, ethyl aluminum dichloride, hsdb 317, dichloro ethyl aluminum
IUPAC Name dichloro(ethyl)alumane
SMILES CC[Al](Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC185451000
SDS View Documents
Thermo Scientific Chemicals
185451000
100 mL AcroSeal™ Glass bottle
Each for $166.13
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Chemical Identifiers

563-43-9
126.94
UAIZDWNSWGTKFZ-UHFFFAOYSA-L
dichloro(ethyl)alumane
C2H5AlCl2
MFCD00000457
ethylaluminum dichloride, aluminum, dichloroethyl, dichloroethylaluminum, ethyldichloroaluminum, dichloromonoethylaluminum, ethylaluminium dichloride, dichloro ethyl alumane, ethyl aluminum dichloride, hsdb 317, dichloro ethyl aluminum
CC[Al](Cl)Cl

Specifications

563-43-9,142-82-5
0.7400g/mL
C2H5AlCl2
MFCD00000457
06,251; 07,146; 10,177
15, 3816
Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons
CC[Al](Cl)Cl
126.94
14.5 to 15.5%
Ethylaluminium dichloride
Colorless
AcroSeal™ Glass bottle
C2H5AlCl2
100 mL
0.74
ethylaluminum dichloride, aluminum, dichloroethyl, dichloroethylaluminum, ethyldichloroaluminum, dichloromonoethylaluminum, ethylaluminium dichloride, dichloro ethyl alumane, ethyl aluminum dichloride, hsdb 317, dichloro ethyl aluminum
UAIZDWNSWGTKFZ-UHFFFAOYSA-L
dichloro(ethyl)alumane
126.95
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effec

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 209-248-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only