Learn More
Ethylaluminium dichloride, 1.8M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 428041000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Ethylaluminium dichloride | |
563-43-9,108-88-3 | |
0.9340g/mL | |
AcroSeal™ Glass bottle | |
C2H5AlCl2 | |
100 mL | |
15, 3816 | |
Solubility in water: reacts | |
CC[Al](Cl)Cl | |
126.94 | |
Liquid |
1.8M solution in toluene | |
Colorless to Yellow | |
4°C | |
C2H5AlCl2 | |
MFCD00000457 | |
0.934 | |
ethylaluminum dichloride, aluminum, dichloroethyl, dichloroethylaluminum, ethyldichloroaluminum, dichloromonoethylaluminum, ethylaluminium dichloride, dichloro ethyl alumane, ethyl aluminum dichloride, hsdb 317, dichloro ethyl aluminum | |
UAIZDWNSWGTKFZ-UHFFFAOYSA-L | |
dichloro(ethyl)alumane | |
126.95 |
Chemical Identifiers
563-43-9 | |
126.94 | |
UAIZDWNSWGTKFZ-UHFFFAOYSA-L | |
dichloro(ethyl)alumane |
C2H5AlCl2 | |
MFCD00000457 | |
ethylaluminum dichloride, aluminum, dichloroethyl, dichloroethylaluminum, ethyldichloroaluminum, dichloromonoethylaluminum, ethylaluminium dichloride, dichloro ethyl alumane, ethyl aluminum dichloride, hsdb 317, dichloro ethyl aluminum | |
CC[Al](Cl)Cl |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May ca
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse
GHS Signal Word: Danger
EINECSNumber : 209-248-6
RUO – Research Use Only