Learn More
Ethylbenzene, 99.8%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
$83.89 - $246.24
Chemical Identifiers
CAS | 100-41-4 |
---|---|
Molecular Formula | C8H10 |
Molecular Weight (g/mol) | 106.17 |
InChI Key | YNQLUTRBYVCPMQ-UHFFFAOYSA-N |
Synonym | phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene |
PubChem CID | 7500 |
ChEBI | CHEBI:16101 |
IUPAC Name | ethylbenzene |
SMILES | CCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC433801000
|
Thermo Scientific Chemicals
433801000 |
100 mL | AcroSeal™ Glass Bottle |
Each for $83.89
|
|
||||
AC433800010
|
Thermo Scientific Chemicals
433800010 |
1 L | AcroSeal™ Glass Bottle |
Each for $246.24
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-41-4 | |
106.17 | |
phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene | |
CHEBI:16101 | |
CCC1=CC=CC=C1 |
C8H10 | |
YNQLUTRBYVCPMQ-UHFFFAOYSA-N | |
7500 | |
ethylbenzene |
Specifications
100-41-4 | |
0.8600g/mL | |
22°C | |
99.75% min. (GC) | |
C8H10 | |
C6H5C2H5 | |
0.0005 % max. | |
15,382 | |
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol,benzene,carbon tetrachloride,,ether and sulfur dioxide,slightly soluble in chloroform,insoluble in ammonia | |
CCC1=CC=CC=C1 | |
106.17 | |
0.63 mPa.s (20°C) | |
106.17 | |
Liquid |
-95.0°C | |
136.0°C | |
Authentic | |
AcroSeal™ Glass Bottle | |
1.4940 to 1.4970 | |
100 mL | |
0.86 | |
phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene | |
YNQLUTRBYVCPMQ-UHFFFAOYSA-N | |
ethylbenzene | |
7500 | |
CHEBI:16101 | |
99.8% | |
Ethylbenzene |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasti
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Danger
EINECSNumber : 202-849-4
RUO – Research Use Only