missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle1-methylcyclopropane-1-carboxylate, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 71441-76-4 |
|---|---|
| Molecular Formula | C7H12O2 |
| Molecular Weight (g/mol) | 128.171 |
| MDL Number | MFCD00001284 |
| InChI Key | IZPYNZLFBUQGCZ-UHFFFAOYSA-N |
| Synonym | ethyl 1-methylcyclopropanecarboxylate, 1-methylcyclopropanecarboxylic acid ethyl ester, ethyl1-methylcyclopropanecarboxylate, cyclopropanecarboxylic acid, 1-methyl-, ethyl ester, cyclopropanecarboxylicacid, 1-methyl-, ethyl ester, ethyl 1-methylcyclopropane-carboxylate, 1-methylcyclopropanoic acid ethyl ester, 1-ethoxycarbonyl-1-methylcyclopropane, ethyl 1-methylcyclopropan-1-carboxylate, ethyl 1-methyl cyclopropane-1-carboxylate |
| PubChem CID | 544234 |
| IUPAC Name | ethyl 1-methylcyclopropane-1-carboxylate |
| SMILES | CCOC(=O)C1(CC1)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
SB01040DA
|
Thermo Scientific Chemicals
SB01040DA |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 71441-76-4 | |
| 128.171 | |
| IZPYNZLFBUQGCZ-UHFFFAOYSA-N | |
| 544234 | |
| CCOC(=O)C1(CC1)C |
| C7H12O2 | |
| MFCD00001284 | |
| ethyl 1-methylcyclopropanecarboxylate, 1-methylcyclopropanecarboxylic acid ethyl ester, ethyl1-methylcyclopropanecarboxylate, cyclopropanecarboxylic acid, 1-methyl-, ethyl ester, cyclopropanecarboxylicacid, 1-methyl-, ethyl ester, ethyl 1-methylcyclopropane-carboxylate, 1-methylcyclopropanoic acid ethyl ester, 1-ethoxycarbonyl-1-methylcyclopropane, ethyl 1-methylcyclopropan-1-carboxylate, ethyl 1-methyl cyclopropane-1-carboxylate | |
| ethyl 1-methylcyclopropane-1-carboxylate |
Specifications
| 71441-76-4 | |
| 0.9180g/mL | |
| Amber Glass Bottle | |
| CH3C3H4C(O)OCH2CH3 | |
| 1g | |
| IZPYNZLFBUQGCZ-UHFFFAOYSA-N | |
| ethyl 1-methylcyclopropane-1-carboxylate | |
| 544234 | |
| 97% |
| 97.0 | |
| 97% | |
| C7H12O2 | |
| MFCD00001284 | |
| ethyl 1-methylcyclopropanecarboxylate, 1-methylcyclopropanecarboxylic acid ethyl ester, ethyl1-methylcyclopropanecarboxylate, cyclopropanecarboxylic acid, 1-methyl-, ethyl ester, cyclopropanecarboxylicacid, 1-methyl-, ethyl ester, ethyl 1-methylcyclopropane-carboxylate, 1-methylcyclopropanoic acid ethyl ester, 1-ethoxycarbonyl-1-methylcyclopropane, ethyl 1-methylcyclopropan-1-carboxylate, ethyl 1-methyl cyclopropane-1-carboxylate | |
| CCOC(=O)C1(CC1)C | |
| 128.171 | |
| 128.17 | |
| Ethyl 1-methylcyclopropane-1-carboxylate |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.