missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ethyle2-(3-methoxyphenyl)acetate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 35553-92-5
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00026897
InChI Key XXVVNHCWPHMLEZ-UHFFFAOYSA-N
Synonym ethyl 3-methoxyphenylacetate, ethyl 2-3-methoxyphenyl acetate, ethyl 3-methoxyphenyl acetate, benzeneacetic acid, 3-methoxy-, ethyl ester, acmc-20akzh, 3-methoxy-phenyl-acetic acid ethyl ester, ethyl 2-3-methoxyphenyl ethanoate, 3-methoxyphenylacetic acid ethyl ester, 3-methoxybenzeneacetic acid ethyl ester, 2-3-methoxyphenyl acetic acid ethyl ester
PubChem CID 2774971
IUPAC Name ethyl 2-(3-methoxyphenyl)acetate
SMILES CCOC(=O)CC1=CC(=CC=C1)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BTB09251DA
SDS View Documents
Thermo Scientific Chemicals
BTB09251DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB09251EA
SDS View Documents
Thermo Scientific Chemicals
BTB09251EA
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB09251EE
SDS View Documents
Thermo Scientific Chemicals
BTB09251EE
50g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

35553-92-5
194.23
XXVVNHCWPHMLEZ-UHFFFAOYSA-N
2774971
CCOC(=O)CC1=CC(=CC=C1)OC
C11H14O3
MFCD00026897
ethyl 3-methoxyphenylacetate, ethyl 2-3-methoxyphenyl acetate, ethyl 3-methoxyphenyl acetate, benzeneacetic acid, 3-methoxy-, ethyl ester, acmc-20akzh, 3-methoxy-phenyl-acetic acid ethyl ester, ethyl 2-3-methoxyphenyl ethanoate, 3-methoxyphenylacetic acid ethyl ester, 3-methoxybenzeneacetic acid ethyl ester, 2-3-methoxyphenyl acetic acid ethyl ester
ethyl 2-(3-methoxyphenyl)acetate

Specifications

35553-92-5
100.0
Amber Glass Bottle
CH3OC6H4CH2C(O)OCH2CH3
1g
XXVVNHCWPHMLEZ-UHFFFAOYSA-N
ethyl 2-(3-methoxyphenyl)acetate
2774971
97%
96.5
97%
C11H14O3
MFCD00026897
ethyl 3-methoxyphenylacetate, ethyl 2-3-methoxyphenyl acetate, ethyl 3-methoxyphenyl acetate, benzeneacetic acid, 3-methoxy-, ethyl ester, acmc-20akzh, 3-methoxy-phenyl-acetic acid ethyl ester, ethyl 2-3-methoxyphenyl ethanoate, 3-methoxyphenylacetic acid ethyl ester, 3-methoxybenzeneacetic acid ethyl ester, 2-3-methoxyphenyl acetic acid ethyl ester
CCOC(=O)CC1=CC(=CC=C1)OC
194.23
194.23
Ethyl 2-(3-methoxyphenyl)acetate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'