missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ethyle2-amino-4-methylpyrimidine-5-carboxylate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CD10070DA

 View more versions of this product

Catalog No. CD10070DA


Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 2-amino-4-methylpyrimidine-5-carboxylate
96.5
97%
C8H11N3O2
1 g
YBFVMJRSZCVJJP-UHFFFAOYSA-N
ethyl 2-amino-4-methylpyrimidine-5-carboxylate
459813
97%
81633-29-6
100.0
Amber Glass Bottle
MFCD00052622
2-amino-4-methylpyrimidine-5-carboxylic acid ethyl ester, 2-amino-4-methyl-pyrimidine-5-carboxylic acid ethyl ester, acmc-209plx, maybridge1_003176, ethyl2-amino-4-methylpyrimidine-5-carboxylate, 2-amino-5-ethoxycarbonyl-4-methylpyrimidine, ethyl 2-amino-4-methyl-pyrimidine-5-carboxylate, ethyl 2-amino-4-methylpyrimidine-5-carboxylate, 2-amino-4-methyl-5-pyrimidinecarboxylic acid, ethyl ester, 5-pyrimidinecarboxylicacid, 2-amino-4-methyl-, ethyl ester
CCOC(=O)C1=CN=C(N)N=C1C
181.20
181.19

Chemical Identifiers

81633-29-6
181.20
YBFVMJRSZCVJJP-UHFFFAOYSA-N
459813
CCOC(=O)C1=CN=C(N)N=C1C
C8H11N3O2
MFCD00052622
2-amino-4-methylpyrimidine-5-carboxylic acid ethyl ester, 2-amino-4-methyl-pyrimidine-5-carboxylic acid ethyl ester, acmc-209plx, maybridge1_003176, ethyl2-amino-4-methylpyrimidine-5-carboxylate, 2-amino-5-ethoxycarbonyl-4-methylpyrimidine, ethyl 2-amino-4-methyl-pyrimidine-5-carboxylate, ethyl 2-amino-4-methylpyrimidine-5-carboxylate, 2-amino-4-methyl-5-pyrimidinecarboxylic acid, ethyl ester, 5-pyrimidinecarboxylicacid, 2-amino-4-methyl-, ethyl ester
ethyl 2-amino-4-methylpyrimidine-5-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions