missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
ethyle7-hydroxy-4-oxo-4H-chromene-2-carboxylate, 97%, Thermo Scientific™
Chemical Identifiers
CAS |
23866-72-0 |
Molecular Formula |
C12H10O5 |
Molecular Weight (g/mol) |
234.21 |
MDL Number |
MFCD00100382 |
InChI Key |
RKIMFICEWCXBCE-UHFFFAOYSA-N |
Synonym |
ethyl 7-hydroxy-4-oxo-4h-chromene-2-carboxylate, 4h-1-benzopyran-2-carboxylic acid, 7-hydroxy-4-oxo-, ethyl ester, chembl82749, 2-ethoxycarbonyl-7-hydroxychromone, ethyl 7-hydroxy-4-oxo-4h-chromen-2-carboxylate, 7-hydroxy-4-oxo-4h-1-benzopyran-2-carboxylic acid ethyl ester, 7-hydroxy-4-oxo-4h-chromene-2-carboxylic acid ethyl ester, 4h-1-benzopyran-2-carboxylicacid, 7-hydroxy-4-oxo-, ethyl ester, maybridge1_002082, 7-hydroxy-4-oxo-4h-chromen-2-carboxylic acid ethyl ester |
PubChem CID |
5376802 |
IUPAC Name |
ethyl 7-hydroxy-4-oxochromene-2-carboxylate |
SMILES |
CCOC(=O)C1=CC(=O)C2=CC=C(O)C=C2O1 |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
23866-72-0 |
|
234.21 |
|
RKIMFICEWCXBCE-UHFFFAOYSA-N |
|
5376802 |
|
CCOC(=O)C1=CC(=O)C2=CC=C(O)C=C2O1 |
|
C12H10O5 |
|
MFCD00100382 |
|
ethyl 7-hydroxy-4-oxo-4h-chromene-2-carboxylate, 4h-1-benzopyran-2-carboxylic acid, 7-hydroxy-4-oxo-, ethyl ester, chembl82749, 2-ethoxycarbonyl-7-hydroxychromone, ethyl 7-hydroxy-4-oxo-4h-chromen-2-carboxylate, 7-hydroxy-4-oxo-4h-1-benzopyran-2-carboxylic acid ethyl ester, 7-hydroxy-4-oxo-4h-chromene-2-carboxylic acid ethyl ester, 4h-1-benzopyran-2-carboxylicacid, 7-hydroxy-4-oxo-, ethyl ester, maybridge1_002082, 7-hydroxy-4-oxo-4h-chromen-2-carboxylic acid ethyl ester |
|
ethyl 7-hydroxy-4-oxochromene-2-carboxylate |
Specifications
|
23866-72-0 |
|
100.0 |
|
Amber Glass Bottle |
|
MFCD00100382 |
|
ethyl 7-hydroxy-4-oxo-4h-chromene-2-carboxylate, 4h-1-benzopyran-2-carboxylic acid, 7-hydroxy-4-oxo-, ethyl ester, chembl82749, 2-ethoxycarbonyl-7-hydroxychromone, ethyl 7-hydroxy-4-oxo-4h-chromen-2-carboxylate, 7-hydroxy-4-oxo-4h-1-benzopyran-2-carboxylic acid ethyl ester, 7-hydroxy-4-oxo-4h-chromene-2-carboxylic acid ethyl ester, 4h-1-benzopyran-2-carboxylicacid, 7-hydroxy-4-oxo-, ethyl ester, maybridge1_002082, 7-hydroxy-4-oxo-4h-chromen-2-carboxylic acid ethyl ester |
|
CCOC(=O)C1=CC(=O)C2=CC=C(O)C=C2O1 |
|
234.21 |
|
234.21 |
|
Ethyl 7-hydroxy-4-oxo-4H-chromene-2-carboxylate |
|
96.5 |
|
97% |
|
C12H10O5 |
|
1g |
|
RKIMFICEWCXBCE-UHFFFAOYSA-N |
|
ethyl 7-hydroxy-4-oxochromene-2-carboxylate |
|
5376802 |
|
97% |
Videos
SDS
missing translation for 'documents'