missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethylene Di(thiotosylate) 98.0+%, TCI America™
Supplier: TCI America E04715G
Specifications
Ethylene Di(thiotosylate) [Protecting Reagent for Active Methylene] | |
74°C | |
C16H18O4S4 | |
5 g | |
DUFUGAKEFZRFEQ-UHFFFAOYSA-N | |
1-methyl-4-[({2-[(4-methylbenzenesulfonyl)sulfanyl]ethyl}sulfanyl)sulfonyl]benzene | |
75212 | |
≥98.0% (HPLC) |
2225-23-2 | |
White | |
MFCD00008550 | |
1,2-Di(p-tosylthio)ethane, 1,2-Ethanedithiol Ditosylate, Ethylene Bis(p-toluenethiosulfonate), S,S′C-Ethylene p-Toluenethiosulfonate | |
CC1=CC=C(C=C1)S(=O)(=O)SCCSS(=O)(=O)C1=CC=C(C)C=C1 | |
402.56 | |
402.56 | |
Crystalline Powder |
Chemical Identifiers
2225-23-2 | |
402.56 | |
DUFUGAKEFZRFEQ-UHFFFAOYSA-N | |
1-methyl-4-[({2-[(4-methylbenzenesulfonyl)sulfanyl]ethyl}sulfanyl)sulfonyl]benzene |
C16H18O4S4 | |
MFCD00008550 | |
75212 | |
CC1=CC=C(C=C1)S(=O)(=O)SCCSS(=O)(=O)C1=CC=C(C)C=C1 |
Safety and Handling
TSCA : No