Learn More
Ethylene diacrylate, 95%, stabilized, Thermo Scientific™
Chemical Identifiers
| CAS | 2274-11-5 |
|---|---|
| Molecular Formula | C8H10O4 |
| Molecular Weight (g/mol) | 170.17 |
| MDL Number | MFCD00008629 |
| InChI Key | KUDUQBURMYMBIJ-UHFFFAOYSA-N |
| Synonym | ethylene diacrylate, ethylene acrylate, ethylene glycol diacrylate, acrylic acid, ethylene ester, acrylic acid, ethylene glycol diester, 2-propenoic acid, 1,2-ethanediyl ester, acrylic acid, diester with ethylene glycol, 1,2-ethanediyl diacrylate, ethylene glycol dipropenoate, unii-42q902riye |
| PubChem CID | 75282 |
| IUPAC Name | 2-prop-2-enoyloxyethyl prop-2-enoate |
| SMILES | C=CC(=O)OCCOC(=O)C=C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC366970050
|
Thermo Scientific Chemicals
366970050 |
5mL | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC366970250
|
Thermo Scientific Chemicals
366970250 |
25mL | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 2274-11-5 | |
| 170.17 | |
| KUDUQBURMYMBIJ-UHFFFAOYSA-N | |
| 75282 | |
| C=CC(=O)OCCOC(=O)C=C |
| C8H10O4 | |
| MFCD00008629 | |
| ethylene diacrylate, ethylene acrylate, ethylene glycol diacrylate, acrylic acid, ethylene ester, acrylic acid, ethylene glycol diester, 2-propenoic acid, 1,2-ethanediyl ester, acrylic acid, diester with ethylene glycol, 1,2-ethanediyl diacrylate, ethylene glycol dipropenoate, unii-42q902riye | |
| 2-prop-2-enoyloxyethyl prop-2-enoate |
Specifications
| 2274-11-5 | |
| 62.0°C to 65.0°C (2.0mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4520 to 1.4560 | |
| 5mL | |
| 1.04 | |
| Solubility in water: slightly soluble | |
| C=CC(=O)OCCOC(=O)C=C | |
| 170.17 | |
| 16790 | |
| 95% |
| 1.0400g/mL | |
| 101°C | |
| 94% min. (GC) | |
| C8H10O4 | |
| MFCD00008629 | |
| 02, 1469 | |
| 2-hydroxy-1-naphthoic acid, 1-naphthalenecarboxylic acid, 2-hydroxy, hydroxynaphthoic acid, 1-naphthoic acid, 2-hydroxy, unii-e3h4qz94uy, e3h4qz94uy, 1-naphthalenecarboxylic acid, hydroxy, 2-hydroxynaphthalenecarboxylic acid, 2-hydroxy-1-naphthoicacid, 2-hydroxynaphtoic acid | |
| KUDUQBURMYMBIJ-UHFFFAOYSA-N | |
| 2-(prop-2-enoyloxy)ethyl prop-2-enoate | |
| 100ppm MEHQ | |
| 170.17 | |
| Ethylene diacrylate, 90% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lastin
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Danger
EINECSNumber : 218-886-4
RUO â Research Use Only