missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethylene Glycol Bis(propionitrile) Ether 97.0+%, TCI America™
Supplier: TCI America E0108100G
Specifications
Ethylene Glycol Bis(propionitrile) Ether | |
Yellow | |
C8H12N2O2 | |
100 g | |
1,2-Bis(2-cyanoethoxy)ethane, 3,3′-(Ethylenedioxy)dipropionitrile, Ethylene Glycol Bis(2-cyanoethyl) Ether | |
C(COCCOCCC#N)C#N | |
168.196 | |
168.20 | |
Liquid |
3386-87-6 | |
165°C | |
MFCD00019871 | |
3276 | |
VTHRQKSLPFJQHN-UHFFFAOYSA-N | |
3-[2-(2-cyanoethoxy)ethoxy]propanenitrile | |
76921 | |
≥97.0% (GC) |
Chemical Identifiers
3386-87-6 | |
168.196 | |
VTHRQKSLPFJQHN-UHFFFAOYSA-N | |
3-[2-(2-cyanoethoxy)ethoxy]propanenitrile |
C8H12N2O2 | |
MFCD00019871 | |
76921 | |
C(COCCOCCC#N)C#N |
Safety and Handling
TSCA : Yes