missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethylene Glycol Dibenzoate 98.0+%, TCI America™
Supplier: TCI America E030025G
Specifications
Ethylene Glycol Dibenzoate | |
70°C | |
C16H14O4 | |
25 g | |
XFDQLDNQZFOAFK-UHFFFAOYSA-N | |
2-(benzoyloxy)ethyl benzoate | |
66750 | |
≥98.0% (GC) |
94-49-5 | |
White | |
MFCD00059685 | |
Glycol Dibenzoate, 1,2-Bis(benzoyloxy)ethane | |
O=C(OCCOC(=O)C1=CC=CC=C1)C1=CC=CC=C1 | |
270.28 | |
270.28 | |
Crystalline Powder |
Chemical Identifiers
94-49-5 | |
270.28 | |
XFDQLDNQZFOAFK-UHFFFAOYSA-N | |
66750 | |
O=C(OCCOC(=O)C1=CC=CC=C1)C1=CC=CC=C1 |
C16H14O4 | |
MFCD00059685 | |
Glycol Dibenzoate, 1,2-Bis(benzoyloxy)ethane | |
2-(benzoyloxy)ethyl benzoate |
Safety and Handling
TSCA : No