missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylene glycol diethyl ether, 99%, Thermo Scientific Chemicals

$133.16 - $504.27

Chemical Identifiers

CAS 629-14-1
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00009253
InChI Key LZDKZFUFMNSQCJ-UHFFFAOYSA-N
Synonym ethylene glycol diethyl ether, diethyl cellosolve, ethane, 1,2-diethoxy, hisolve eme, 3,6-dioxaoctane, glyme-1, diethyl glycol, 2-ethoxyethyl ethyl ether, egdee, diethylether ethylenglykolu
PubChem CID 12375
IUPAC Name 1,2-diethoxyethane
SMILES CCOCCOCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC208241000
SDS View Documents
Thermo Scientific Chemicals
208241000
100 mL Glass bottle
Each for $133.16
 
AC208240010
SDS View Documents
Thermo Scientific Chemicals
208240010
1 L Glass bottle
Each for $504.27
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

629-14-1
118.18
LZDKZFUFMNSQCJ-UHFFFAOYSA-N
12375
CCOCCOCC
C6H14O2
MFCD00009253
ethylene glycol diethyl ether, diethyl cellosolve, ethane, 1,2-diethoxy, hisolve eme, 3,6-dioxaoctane, glyme-1, diethyl glycol, 2-ethoxyethyl ethyl ether, egdee, diethylether ethylenglykolu
1,2-diethoxyethane

Specifications

629-14-1
Colorless
121.0°C
Authentic
Glass bottle
1.3910 to 1.3930
MFCD00009253
01, 468
ethylene glycol diethyl ether, diethyl cellosolve, ethane, 1,2-diethoxy, hisolve eme, 3,6-dioxaoctane, glyme-1, diethyl glycol, 2-ethoxyethyl ethyl ether, egdee, diethylether ethylenglykolu
LZDKZFUFMNSQCJ-UHFFFAOYSA-N
1,2-diethoxyethane
12375
118.18
Liquid
-74.0°C
0.8417g/mL
19°C
98% min. (GC)
C6H14O2
C2H5OCH2CH2OC2H5
100 mL
0.8417
Solubility in water: 20.4%.
CCOCCOCC
118.18
0.7 mPa.s (20°C)
99%
Ethylene glycol diethyl ether
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May damage the unborn child.
Suspected of damaging fertility.
Highly flammable liquid and vapor.
May form explosive peroxides.

GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES

GHS Signal Word: Danger

EINECSNumber : 211-076-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only