missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethylene Glycol Mono-tert-butyl Ether 99.0+%, TCI America™
Supplier: TCI America E0354500ML
Specifications
Ethylene Glycol Mono-tert-butyl Ether | |
Colorless | |
C6H14O2 | |
500 mL | |
2-tert-Butoxyethanol, tert-Butyl Cellosolve | |
CC(C)(C)OCCO | |
118.18 | |
118.18 | |
Liquid |
7580-85-0 | |
152°C | |
MFCD00059004 | |
3271 | |
BDLXTDLGTWNUFM-UHFFFAOYSA-N | |
2-(tert-butoxy)ethan-1-ol | |
24232 | |
≥99.0% (GC) |
Chemical Identifiers
7580-85-0 | |
118.18 | |
BDLXTDLGTWNUFM-UHFFFAOYSA-N | |
24232 | |
CC(C)(C)OCCO |
C6H14O2 | |
MFCD00059004 | |
2-tert-Butoxyethanol, tert-Butyl Cellosolve | |
2-(tert-butoxy)ethan-1-ol |
Safety and Handling
EINECSNumber : (2)-2424&(7)-0097
RTECSNumber : KJ8750000
TSCA : No