missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals

$59.52 - $651.95

Chemical Identifiers

CAS 333-18-6
Molecular Formula C2H10Cl2N2
Molecular Weight (g/mol) 133.02
MDL Number MFCD00012524
InChI Key OHHBFEVZJLBKEH-UHFFFAOYSA-N
Synonym ethylenediamine dihydrochloride, chlor-ethamine, ethylenediammonium chloride, ethylenediammonium dichloride, ethane-1,2-diamine dihydrochloride, dimethylenediamine dihydrochloride, 1,2-diaminoethane dihydrochloride, 1,2-ethanediamine, dihydrochloride, ethylenediamine 2hcl, dimethylenediamine diydrochloride
PubChem CID 9521
ChEBI CHEBI:53626
IUPAC Name ethane-1,2-diamine;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1356322
View Documents
Thermo Scientific Chemicals
A1356322
100 g
Each for $59.52
 
AAA1356336
View Documents
Thermo Scientific Chemicals
A1356336
500 g
Each for $184.44
 
AAA135630E
View Documents
Thermo Scientific Chemicals
A135630E
2500 g
Each for $651.95
 
Description

Description

Ethylenediamine dihydrochloride was used in modified ethylenediamine condensation method for fluorimetric determination of catecholamines. It was used to investigate the luminescence properties of complexes of EuIII and TbIII with ethylenediamine. It was used in the synthesis of 1,3,5-tris(4,5-dihydro-1H-imidazol-2-yl)benzene3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediamine dihydrochloride was used in modified ethylenediamine condensation method for fluorimetric determination of catecholamines. It was used to investigate the luminescence properties of complexes of EuIII and TbIII with ethylenediamine. It was used in the synthesis of 1,3,5-tris(4,5-dihydro-1H-imidazol-2-yl)benzene3

Solubility
Soluble in water 300 g/L (20°C).

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

333-18-6
133.02
OHHBFEVZJLBKEH-UHFFFAOYSA-N
9521
ethane-1,2-diamine;dihydrochloride
C2H10Cl2N2
MFCD00012524
ethylenediamine dihydrochloride, chlor-ethamine, ethylenediammonium chloride, ethylenediammonium dichloride, ethane-1,2-diamine dihydrochloride, dimethylenediamine dihydrochloride, 1,2-diaminoethane dihydrochloride, 1,2-ethanediamine, dihydrochloride, ethylenediamine 2hcl, dimethylenediamine diydrochloride
CHEBI:53626
[H+].[H+].[Cl-].[Cl-].NCCN

Specifications

333-18-6
C2H10Cl2N2
100 g
ethylenediamine dihydrochloride, chlor-ethamine, ethylenediammonium chloride, ethylenediammonium dichloride, ethane-1,2-diamine dihydrochloride, dimethylenediamine dihydrochloride, 1,2-diaminoethane dihydrochloride, 1,2-ethanediamine, dihydrochloride, ethylenediamine 2hcl, dimethylenediamine diydrochloride
OHHBFEVZJLBKEH-UHFFFAOYSA-N
ethane-1,2-diamine;dihydrochloride
9521
133.02
Ethylenediamine dihydrochloride
>300°C
MFCD00012524
Hygroscopic
Soluble in water 300g/L (20°C).
[H+].[H+].[Cl-].[Cl-].NCCN
133.02
CHEBI:53626
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H373-H302
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.

P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P341-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c

H302-H315-H317-H319-H334-H335

EINECSNumber : 206-369-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only