missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethylenediamine Sulfate 98.0+%, TCI America™
$61.38 - $305.25
Chemical Identifiers
CAS | 22029-36-3 |
---|---|
Molecular Formula | C2H10N2O4S |
Molecular Weight (g/mol) | 158.17 |
MDL Number | MFCD00036417 |
InChI Key | BNZCDZDLTIHJAC-UHFFFAOYSA-N |
PubChem CID | 168015 |
IUPAC Name | ethane-1,2-diamine; sulfuric acid |
SMILES | NCCN.OS(O)(=O)=O |
Chemical Identifiers
22029-36-3 | |
158.17 | |
BNZCDZDLTIHJAC-UHFFFAOYSA-N | |
ethane-1,2-diamine; sulfuric acid |
C2H10N2O4S | |
MFCD00036417 | |
168015 | |
NCCN.OS(O)(=O)=O |
Specifications
22029-36-3 | |
C2H10N2O4S | |
25 g | |
BNZCDZDLTIHJAC-UHFFFAOYSA-N | |
ethane-1,2-diamine; sulfuric acid | |
168015 | |
≥98.0% (T) | |
Ethylenediamine Sulfate |
White | |
MFCD00036417 | |
2811 | |
NCCN.OS(O)(=O)=O | |
158.17 | |
158.17 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0150
RTECSNumber : KV5425000
TSCA : Yes