missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0), Fisher BioReagents

Supplier:  Fisher BioReagents BP248220

 View more versions of this product

Catalog No. BP248220


Explore available promotions

Description

Description

EDTA is used extensively in molecular biology to minimize metal ion impurities in reaction buffers. Ideal for DNA work.
Specifications

Specifications

Ethylenediamine Tetraacetic Acid
0.5 M Solution, pH 8.0 (for DNA Work)
C10H16N2O8
MFCD00003541
15, 3565
KCXVZYZYPLLWCC-UHFFFAOYSA-N
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
6049
292.25
Poly CUBE
20 L
Colorless
60-00-4,7732-18-5
(HOOCCH2)2NCH2CH2N(CH2COOH)2
0.475 to 0.525M
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
292.24
CHEBI:42191
Pass Test
8.0
Liquid

Chemical Identifiers

60-00-4
292.24
KCXVZYZYPLLWCC-UHFFFAOYSA-N
6049
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
C10H16N2O8
MFCD00003541
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
CHEBI:42191
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS Documents

Documents

Promotions

Promotions