missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediaminetetraacetic acid disodium salt, 0.200N (0.1M) Standardized solution, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 039783K2

 View more versions of this product

Catalog No. AA39783K2


Explore available promotions

Description

Description

Ethylenediaminetetraacetic acid disodium salt is widely used in textile industry. Usually applied to dissolve limescale. It is applied in textile industry, pulp and paper industry and also in chelation therapy. In cosmetics, it acts as a sequestering agent. It acts as a corrosion inhibitor to carbon steel in the industries. It also acts as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethylenediaminetetraacetic acid disodium salt
Colorless
Odorless
MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
ZGTMUACCHSMWAC-UHFFFAOYSA-L
disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
57339238
336.22
139-33-3
0.200N (0.1M)
C10H14N2Na2O8
1 L
Miscible with water.
[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
336.21
CHEBI:64734
Liquid

Chemical Identifiers

139-33-3
336.21
ZGTMUACCHSMWAC-UHFFFAOYSA-L
57339238
disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
C10H14N2Na2O8
MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
CHEBI:64734
[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-358-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only