missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate, ≥99%, Molecular Biology Reagent Grade, MP Biomedicals™
$69.01 - $370.35
Identifiants chimiques
CAS | 6381-92-6 |
---|---|
Molecular Formula | C10H18N2Na2O10 |
Molecular Weight (g/mol) | 372.24 |
MDL Number | MFCD00150037,MFCD00003541 |
InChI Key | OVBJJZOQPCKUOR-UHFFFAOYSA-L |
Synonym | edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs |
PubChem CID | 44120005 |
SMILES | O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
ICN19482250
|
MP Biomedicals Inc
0219482250 |
50 g |
chaque for $69.01
|
|
|||||
ICN19482280
|
MP Biomedicals Inc
0219482280 |
100 g |
chaque for $104.90
|
|
|||||
ICN19482283
|
MP Biomedicals Inc
0219482283 |
250 g |
chaque for $198.14
|
|
|||||
ICN19482290
|
MP Biomedicals Inc
0219482290 |
500 g |
chaque for $298.36
|
|
|||||
ICN19482291
|
MP Biomedicals Inc
0219482291 |
1 kg |
chaque for $370.35
|
|
|||||
Identifiants chimiques
6381-92-6 | |
372.24 | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
44120005 |
C10H18N2Na2O10 | |
MFCD00150037,MFCD00003541 | |
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
Spécifications
6381-92-6 | |
≥99% | |
MFCD00150037,MFCD00003541 | |
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
372.24 | |
372.2 | |
Crystalline Powder |
White | |
C10H18N2Na2O10 | |
50 g | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate | |
44120005 | |
Molecular Biology Reagent | |
Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate |
Sécurité et manipulation
missing translation for 'einecsNumber' : 205-358-3
Recommended Storage : Store at Room Temperature (15 to 30°C).