Learn More
Ethylenediaminetetraacetic acid disodium salt dihydrate, 99+%, Thermo Scientific Chemicals
$94.89 - $1022.66
Chemical Identifiers
CAS | 6381-92-6 |
---|---|
Molecular Formula | C10H18N2Na2O10 |
Molecular Weight (g/mol) | 372.24 |
MDL Number | MFCD00150037,MFCD00003541 |
InChI Key | OVBJJZOQPCKUOR-UHFFFAOYSA-L |
Synonym | edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs |
PubChem CID | 44120005 |
IUPAC Name | disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate |
SMILES | O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1516130
|
Thermo Scientific Chemicals
A1516130 |
250 g |
Each for $94.89
|
|
|||||
AAA151610B
|
Thermo Scientific Chemicals
A151610B |
1000 g |
Each for $262.58
|
|
|||||
AAA151610I
|
Thermo Scientific Chemicals
A151610I |
5000 g |
Each for $1,022.66
|
|
|||||
Description
Ethylenediaminetetraacetic acid disodium salt dihydrate is used as a chelator of divalent cations. It inhibits enzymes such as metalloproteases that require divalent cations for activity. It is also used in chelation therapy. It acts as a sequestering agent in shampoos, cleaners and other personal care products and serves as an anticoagulant for blood samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthylenediaminetetraacetic acid disodium salt dihydrate is used as a chelator of divalent cations. It inhibits enzymes such as metalloproteases that require divalent cations for activity. It is also used in chelation therapy. It acts as a sequestering agent in shampoos, cleaners and other personal care products and serves as an anticoagulant for blood samples.
Solubility
Soluble in water.
Notes
Hygroscopic.Incompatible with strong oxidizing agents, strong bases, metals and copper.
Chemical Identifiers
6381-92-6 | |
372.24 | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
44120005 | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
C10H18N2Na2O10 | |
MFCD00150037,MFCD00003541 | |
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate |
Specifications
6381-92-6 | |
C10H18N2Na2O10 | |
250 g | |
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate | |
44120005 | |
≥99% |
248°C (decomposition) | |
MFCD00150037,MFCD00003541 | |
3900609 | |
Soluble in water. | |
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
372.24 | |
372.24 (336.22 Anhydrous) | |
Ethylenediaminetetraacetic acid disodium salt dihydrate |
Safety and Handling
P260-P271-P304+P340-P312-P314-P501c
H332-H373
EINECSNumber : 205-358-3
RTECSNumber : AH4375000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only