missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylenediaminetetraacetic acid zinc disodium salt hydrate, Thermo Scientific Chemicals

$70.14 - $614.92

Chemical Identifiers

CAS 14025-21-9
Molecular Formula C10H12N2Na2O8Zn
MDL Number MFCD00135187
InChI Key UBLOJEHIINPTTG-UHFFFAOYSA-J
Synonym zinc disodium edta, edta disodium zinc salt, unii-r84ov391ba, sequestrene na2zn zinc chelate, disodium zinc edetate, disodium zinc ethylenediaminetetraacetate, zinc disodium ethylenediaminetetraacetate, disodium zinc ethylenediaminetetraacetate dihydrate, acetic acid, ethylenedinitrilo tetra-, disodium salt, zinc complex, dihydrate, ethylenediaminetetraacetic acid disodium zinc salt
PubChem CID 51808
IUPAC Name disodium;zinc;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA2242614
View Documents
Thermo Scientific Chemicals
02242614
25 g
Each for $70.14
 
AA2242622
View Documents
Thermo Scientific Chemicals
02242622
100 g
Each for $173.90
 
AA2242636
View Documents
Thermo Scientific Chemicals
02242636
500 g
Each for $614.92
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14025-21-9
MFCD00135187
zinc disodium edta, edta disodium zinc salt, unii-r84ov391ba, sequestrene na2zn zinc chelate, disodium zinc edetate, disodium zinc ethylenediaminetetraacetate, zinc disodium ethylenediaminetetraacetate, disodium zinc ethylenediaminetetraacetate dihydrate, acetic acid, ethylenedinitrilo tetra-, disodium salt, zinc complex, dihydrate, ethylenediaminetetraacetic acid disodium zinc salt
disodium;zinc;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate
C10H12N2Na2O8Zn
UBLOJEHIINPTTG-UHFFFAOYSA-J
51808

Specifications

14025-21-9
MFCD00135187
zinc disodium edta, edta disodium zinc salt, unii-r84ov391ba, sequestrene na2zn zinc chelate, disodium zinc edetate, disodium zinc ethylenediaminetetraacetate, zinc disodium ethylenediaminetetraacetate, disodium zinc ethylenediaminetetraacetate dihydrate, acetic acid, ethylenedinitrilo tetra-, disodium salt, zinc complex, dihydrate, ethylenediaminetetraacetic acid disodium zinc salt
[Na+].[Na+].[Zn++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
399.57
399.50 (Anhydrous)
Ethylenediaminetetraacetic acid zinc disodium salt hydrate
C10H12N2Na2O8Zn
25 g
UBLOJEHIINPTTG-UHFFFAOYSA-J
disodium;zinc;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate
51808
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 237-865-0

RTECSNumber : ZG9213000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only