missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Etoposide, Thermo Scientific Chemicals

Topoisomerase inhibitor and inducer of apoptosis

Supplier:  Thermo Scientific Chemicals J63651MB

 View more versions of this product

Catalog No. AAJ63651MB


Explore available promotions

Description

Description

Etoposide is used as a inhibits DNA synthesis and is very active against cells in the late S and G2 phases of the cell cycle. Etoposide was reported to be used to serum starve Saos-2 M5(p53-/-) cells which were transfected by electroporation with indicated expression plasmids for flag-E2F1 and myc-EBNA3C for selection of colony forming units, thus studying the blocking effect of EBV Nuclear Antigen 3C on E2F1 Mediated Apoptosis Induced by the DNA Damage Response.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Etoposide is used as a inhibits DNA synthesis and is very active against cells in the late S and G2 phases of the cell cycle. Etoposide was reported to be used to serum starve Saos-2 M5(p53-/-) cells which were transfected by electroporation with indicated expression plasmids for flag-E2F1 and myc-EBNA3C for selection of colony forming units, thus studying the blocking effect of EBV Nuclear Antigen 3C on E2F1 Mediated Apoptosis Induced by the DNA Damage Response.

Solubility
Insoluble in water.

Notes
Store solutions in DMSO at 4°C. For long term storage prepare aliquots and store at -20°C.
Specifications

Specifications

Etoposide
236°C to 251°C
C29H32O13
25 mg
vjjpusntgommgy-nzlmilqcsa
VJJPUSNTGOMMGY-MRVIYFEKSA-N
COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12
588.56
588.56
33419-42-0
White
MFCD00869325,MFCD00869325
14,3886
Insoluble in water.
−107° (c=1 in Methanol/Dichloromethane 1:9)
(10R,11R,15R,16S)-16-{[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one
50936917
Powder

Chemical Identifiers

33419-42-0
588.56
VJJPUSNTGOMMGY-MRVIYFEKSA-N
50936917
COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12
C29H32O13
MFCD00869325,MFCD00869325
vjjpusntgommgy-nzlmilqcsa
(10R,11R,15R,16S)-16-{[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H302
May cause cancer.
Harmful if swallowed.

P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H350

EINECSNumber : 251-509-1

RTECSNumber : KC0190000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only