missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eugenol, 99%, Thermo Scientific Chemicals

$73.91 - $1217.07

Chemical Identifiers

CAS 97-53-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008654
InChI Key RRAFCDWBNXTKKO-UHFFFAOYSA-N
Synonym eugenol, 4-allylguaiacol, 4-allyl-2-methoxyphenol, eugenic acid, allylguaiacol, caryophyllic acid, p-allylguaiacol, p-eugenol, 2-methoxy-4-allylphenol, engenol
PubChem CID 3314
ChEBI CHEBI:4917
SMILES COC1=CC(CC=C)=CC=C1O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC119110050
SDS View Documents
Thermo Scientific Chemicals
119110050
5 g Glass bottle
Each for $73.91
 
AC119111000
SDS View Documents
Thermo Scientific Chemicals
119111000
100 g Glass bottle
Each for $86.86
 
AC119115000
SDS View Documents
Thermo Scientific Chemicals
119115000
500 g Glass bottle
Each for $290.42
 
AC119110025
SDS View Documents
Thermo Scientific Chemicals
119110025
2.5 kg Glass bottle
Each for $1,217.07
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

97-53-0
164.20
RRAFCDWBNXTKKO-UHFFFAOYSA-N
3314
COC1=CC(CC=C)=CC=C1O
C10H12O2
MFCD00008654
eugenol, 4-allylguaiacol, 4-allyl-2-methoxyphenol, eugenic acid, allylguaiacol, caryophyllic acid, p-allylguaiacol, p-eugenol, 2-methoxy-4-allylphenol, engenol
CHEBI:4917

Specifications

97-53-0
Yellow
254.0°C
Authentic
Glass bottle
1.5400 to 1.5420
MFCD00008654
06,961
15,394
Solubility in water: 2.46g/L (25°C). Other solubilities: soluble in glacial acetic acid,soluble in alcohol,chloroform and ether
COC1=CC(CC=C)=CC=C1O
164.20
CHEBI:4917
99%
Eugenol, 99%
-12°C to 10°C
1.0600g/mL
>112°C
98.5% min. (GC)
C10H12O2
(H2C=CHCH2)C6H3(OCH3)OH
5 g
1.06
eugenol, 4-allylguaiacol, 4-allyl-2-methoxyphenol, eugenic acid, allylguaiacol, caryophyllic acid, p-allylguaiacol, p-eugenol, 2-methoxy-4-allylphenol, engenol
RRAFCDWBNXTKKO-UHFFFAOYSA-N
2-methoxy-4-prop-2-enylphenol
3314
164.2
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice

GHS Signal Word: Warning

EINECSNumber : 202-589-1

RTECSNumber : SJ4375000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only