Globe Scientific Euromex HD-Mini Camera
This HD-Mini camera with tablet is the perfect solution for modern microscopy in industrial and laboratory applications where real time images are needed. This HD image quality camera can be used for biological, metallurgical or stereo microscopes.
Supplier: Globe Scientific EVC3024HDS
Description
- High definition 1080P color camera
- C-mount interface with c-mount lens (23.2) included
- HDMI and SD memory card
- Real time images directly on screen
- Built-in mouse-controlled software
- Wide array of measurement functions
- 13 in. HD screen included
STAND-ALONE CAPABILITIES
- No need for a computer or any computer knowledge to use this camera. Most camera settings are performed automatically for user friendly operation.
- Simply connect the camera to a high-definition HD screen, TV-HD or HD-beamer with an HDMI input and the system is ready to go.
- The mouse-driven built-in software of the cameras enables capturing 1920 x 1080 pixels jpg images on the integrated SD memory card.
- The embedded software of the camera features functions including:
- image and video capture
- white balance
- auto/manual exposure
- zoom in/out
- horizontal/vertical flip
- video freeze/cancel video freeze
- grid
- display/hide camera control panel
- color adjustment: sharpness/de-noise/saturation/gamma/contrast
- wide array of measurement functions
- These key features make the camera an ideal choice for real time inspections, educational purposes and live demonstrations.
Specifications
Camera | |
Not Autoclavable | |
Non-sterile | |
110/220 V |