Learn More
Europium(III) oxide, 99.9% (REO), Thermo Scientific Chemicals
For vacuum deposition
Supplier: Thermo Scientific Chemicals 03635204
Description
Europium(III) oxide is used for vacuum deposition, fluorescent lamps, and as an activator for yttrium-based phosphors, manufacture of fluorescent glass, anti-counterfeiting phosphors in Euro banknotes. And it is widely used as a red or blue phosphor in television sets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Europium(III) oxide | |
1308-96-9 | |
2 g | |
Eu2O3 | |
14,3904 | |
Soluble in acids. Insoluble in water. | |
[O--].[O--].[O--].[Eu+3].[Eu+3] | |
351.93 | |
351.92 | |
Hygroscopic |
2291°C | |
Sintered Lump | |
(REO) | |
MFCD00010997 | |
europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2- | |
AEBZCFFCDTZXHP-UHFFFAOYSA-N | |
dieuropium(3+) trioxidandiide | |
159371 | |
99.9% |
Chemical Identifiers
1308-96-9 | |
351.93 | |
AEBZCFFCDTZXHP-UHFFFAOYSA-N | |
159371 |
Eu2O3 | |
MFCD00010997 | |
europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2- | |
[O--].[O--].[O--].[Eu+3].[Eu+3] |
Safety and Handling
EINECSNumber : 215-165-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only