missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Europium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 04030509

Catalog No. AA4030509


Explore available promotions

Description

Description

It is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Europium(III) trifluoromethanesulfonate
52093-25-1
10 g
Eu(OSO2CF3)3
europium trifluoromethanesulfonate, europium iii trifluoromethanesulfonate, europium iii triflate, europium triflate, eu otf 3, ksc268k4f, europium 3+ tritriflate, europium iii tris trifluoromethanesulfonate, europium 3+ ion tritrifluoromethanesulfonate
TWNOVENTEPVGEJ-UHFFFAOYSA-K
europium(3+);trifluoromethanesulfonate
9960317
98%
White
Powder
C3EuF9O9S3
MFCD00209587
Insoluble in water.
[Eu+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
599.15
599.15
Hygroscopic

Chemical Identifiers

52093-25-1
599.15
TWNOVENTEPVGEJ-UHFFFAOYSA-K
9960317
[Eu+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
C3EuF9O9S3
MFCD00209587
europium trifluoromethanesulfonate, europium iii trifluoromethanesulfonate, europium iii triflate, europium triflate, eu otf 3, ksc268k4f, europium 3+ tritriflate, europium iii tris trifluoromethanesulfonate, europium 3+ ion tritrifluoromethanesulfonate
europium(3+);trifluoromethanesulfonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only