Fisherbrand™ EVA Foam Ice Pans and Buckets with Lids
Fisherbrand™EVA Foam Ice Pans and Buckets with Lids provide excellent insulation for any benchtop cryogenic application, without the mess!
 View more versions of this product
Catalog No. 03395161
Explore available promotions
Includes: removable lid.
Description
Description
  • Sweat-proof ethylene vinyl acetate will not leak
  • Compatible with ice, dry ice, liquid nitrogen, alcohol, and saline solutions
  • Bases and lids feature glove-friendly handles
  • Cleanup is quick and easy with molded pour spouts
Specifications
Specifications
Ice, dry ice, liquid nitrogen, alcohol, and saline solutions
Bucket with Lid
Cryogenic Storage Box
Purple
Not Autoclavable
Removable lid
EVA
1
2.5 L