Learn More
Caplugs™ Evergreen Scientific CryoSure™ Brand Cryogenic Vials with Caps
External thread, freestanding vials with caps provide safe, secure, reliable storage for specimens in liquid nitrogen vapor-phase™
$489.76 - $601.79
Specifications
Closure Color | Natural |
---|---|
Diameter (English) Outer | 0.49 in. |
Color | Clear |
Diameter (Metric) Outer | 12.5 mm |
Material | Polypropylene |
Catalog Number | Mfr. No. | Volume (Metric) | Height (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Height (Metric) | Price | Quantity | |||||
05-557-0
|
Caplugs™
2223901085 |
1 mL | 41 mm |
Case of 500 Each for $601.79
|
|
|||||
05-554-5
|
Caplugs™
2223905085 |
3.5 mL | 76 mm |
Case of 500 Each for $558.33
|
|
|||||
05-556-4
|
Caplugs™
2223902085 |
1.5 mL | 46 mm |
Case of 500 Each for $489.76
|
|
|||||
Description
Recommended for storing cell cultures, blood/serum specimens, sperm, and other biological specimens at vapor phase™ liquid nitrogen temperatures (-320°F/-195°C).
- External thread design is less susceptible to aerosol contamination than internally threaded vials, and have smoother engagement than ordinary 60° threads
- Ultra-clear polypropylene material is nearly transparent and provides superior tensile strength and resistance to gamma radiation sterilization
- Permanent marking areas are molded in for labeling and specimen identification, with volume graduations distinctly molded every 0.5mL
- Straight vertical inside walls with no shoulder result in a large accessible opening which makes processing and retrieval of samples convenient
Polyethylene Caps – Natural Color
- Provide leakproof closure without the use of a silicone gasket
- Feature a molded-in sealing ring for a rim-seal that offers sealing efficacy in excess of 99%
Specifications
Natural | |
Clear | |
Polypropylene | |
Vapor phase liquid nitrogen only | |
Screw Cap |
0.49 in. | |
12.5 mm | |
Sterile | |
External | |
500/Cs. |
™Please use CryoSure™ vials in the vapor-phase only. Evergreen does not recommend the use of screwcap vials, including CryoSure™ vials, for immersion in liquid nitrogen or other liquified gases. Immersion in liquid nitrogen may cause leakage in CryoSure™ vials and also presents the potential danger of rapid pressure buildup and/or explosion when returned to room temperature.