Caplugs™ Evergreen Scientific Dilution Vials in Tray
Dilution vials provide accurate, hemolysis- and distortion-free cell and particle counts
Supplier:  Caplugs™ 201526605SF
Catalog No. 05-551-3
Explore available promotions
Description
Description
  • One-piece polystyrene design
  • With rounded seams, frosted writing area and optically clear, parallel sides for optimum light transmission
  • Leakproof caps have convenient tabs for easy removal
  • Vials manufactured and packed in a dust- and particle-free environment; rigorous quality control protocols ensure consistent cleanliness
  • 5 trays of 50
Specifications
Specifications
Polystyrene
Accurate, hemolysis and distortion free cell and particle counts
250 Cs.
Non-sterile
Rounded seams, a frosted writing area and an optically clear parallel sides for optimum light transmission