Caplugs™ (Evergreen Scientific) Pap-Fix Plus™ Empty Pap Jars
Deliver outstanding performance in transporting Pap smear slides
Supplier:  Caplugs™ (Evergreen Scientific) 2405400K8K
 View more versions of this product
Catalog No. 05-557-2
Explore available promotions
Description
Description
  • Rigid polypropylene construction provides fortress-like protection for up to four slides in transit
  • Pap jars shipped empty in bulk packs
Specifications
Specifications
Pap Jar
Fuchsia
Preserving and transporting pap smear slides
Polypropylene
Cap
1000 Cs.