Caplugs™ (Evergreen Scientific) Polyethylene Stopper Caps
Fit test tubes, culture tubes and sample cups for liquid handling
$83.97 - $329.38
Specifications
Material Polyethylene
View More Specs
Products 12
Catalog Number Mfr. No. Color Product Type For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Color Product Type For Use With (Equipment) Price Quantity  
05-555-15
View Documents
Caplugs™ (Evergreen Scientific)
3001999020
Natural Pierceable Stopper Automated Analyzer Sample Cups
Pack of 1000 Each for $124.22
05-551-8
View Documents
Caplugs™ (Evergreen Scientific)
3002912G20
Green Stopper Cap 12 mm Tubes
Pack of 1000 Each for $116.97
05-555-16
View Documents
Caplugs™ (Evergreen Scientific)
3002912W20
Natural Stopper Cap 12 mm Tubes
Pack of 1000 Each for $116.97
05-551-4
View Documents
Caplugs™ (Evergreen Scientific)
3002916R20
Red Stopper Cap 16 mm Tubes
Pack of 1000 Each for $329.38
05-300-291
View Documents
Caplugs™ (Evergreen Scientific)
3002913Y20
Yellow Stopper Cap 13 mm Tubes
Case of 1000 Each for $161.00
05-555-19
View Documents
Caplugs™ (Evergreen Scientific)
300381202P
Natural 2-position Stopper Cap 12 mm Culture Tubes
Case of 1000 Each for $167.53
05-555-17
View Documents
Caplugs™ (Evergreen Scientific)
3002916020
Natural Stopper Cap 16 mm Tubes
Pack of 1000 Each for $250.00
05-555-20
View Documents
Caplugs™ (Evergreen Scientific)
300381202S
Natural 2-position Stopper Cap 12 mm Culture Tubes
Pack of 1000 Each for $240.63
05-552-9
View Documents
Caplugs™ (Evergreen Scientific)
300471702N
Natural Stopper Cap 17 mm Tubes
Case of 1000 Each for $188.94
05-540-2
View Documents
Caplugs™ (Evergreen Scientific)
3002917B2N
Blue Stopper Cap 17 mm Tubes
Pack of 1000 Each for $190.53
05-558-34
View Documents
Caplugs™ (Evergreen Scientific)
3004710020
Natural Flat Top Stopper Cap 10 mm Tubes
Case of 1000 Each for $215.00
02-544-73A
View Documents
Caplugs™ (Evergreen Scientific)
3001006020
Natural Stopper Cap Automated Analyzer Sample Cups
Pack of 1000 Each for $83.97
Description
Description
For test tubes:
  • Precision-molded stopper caps are available for every size (except 15.5 × 50mm) of Evergreen test tube.
  • Choose from either standard caps (like the 300-2900 series) or the flat-top style (300-4700 series).
For culture tubes:
  • Over-fitting caps for use with sterile culture tubes have two positions: loose for aerobic culturing; tight for anaerobic culturing and leak-proof sealing.
  • Caps for culture tubes are gamma-irradiated for sterility.
For sample cups:
  • Perforated caps (Cat. No. 05-551-5) help minimize variation caused by evaporation during processing; non-perforated caps and push-button stoppers are appropriate for short-term storage.(Cat. Nos. 02-544-73A and 05-300-291)
  • See Evergreen Scientific sample cups
Specifications
Specifications
Polyethylene
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'