Invitrogen™ EVOS™ M7000 Imaging System Extended Warranty Package
The EVOS M7000 Imaging System is a high-performance, fully automated, inverted, multi-channel fluorescence and transmitted light imaging system. It was developed to meet today’s expectations for image quality, user interface interactions, data generation speed and flexibility, and end-user value.
Supplier:  Invitrogen™ A42844
Catalog No. A42844
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
This package includes the EVOS M7000 Imaging System, SmartStart orientation, and an extended warranty.
The EVOS M7000 Imaging System Extended Warranty Package offers these advantages:
  • Outstanding image quality and versatility with a 5-position objective turret, 4-color LED fluorescence and transmitted light channels, and 3.2 MP CMOS color and B/W cameras
  • Exceptional usability with fully automated X/Y scanning stage, autofocus, and acquisition routines
  • High-speed image acquisition coupled with multi-position well scanning, and Z-stack and tile-stitch options lend power and flexibility to your data generation
  • Fully integrated time-lapse live cell imaging using the optional EVOS Onstage Incubator for precise control of temperature, gases for normoxic or hypoxic conditions, and humidity
  • Powerful image analysis capabilities for cell segmentation and quantification with the optional Celleste™ Image Analysis Software
  • 1-year extended warranty including one planned maintenance visit
Service plans help protect your investment
  • Maximize system uptime with fast repair turnaround time from manufacturer-trained and certified field service engineers
  • Reduce overall repair costs with remote instrument diagnosis and resolution
Get up and running quickly with SmartStart
The EVOS M7000 Imaging System Extended Warranty Package includes one day of on-site SmartStart orientation carried out by a Field Application Scientist to get you up and running quickly in your lab and reduce downtime. SmartStart is the first step of your education journey and is designed to help you work smarter and with greater efficiency. It includes:
  • Basic setup
  • Review of hardware and fluidics
  • Instrument operation and maintenance
  • Software use
  • Sample preparation
  • Applications
  • Data analysis
  • Troubleshooting
Content will vary depending on your level of knowledge and experience.
Fully automated imaging system
The EVOS M7000 system combines and integrates precision components with a unique modern design functionality that enables high-quality automated fluorescence imaging anywhere with unprecedented workflow efficiency. Full automation of the precision X/Y-stage movement; changing of the 4 LED fluorescent light cubes, 5-position objective turret, focus, and exposure; and switching of the dual camera make the EVOS M7000 system an exceptional platform for a variety of demanding imaging applications. Through the intuitive acquisition interface, you can program the EVOS M7000 system to run well-plate scanning, time lapse experiments, and tile-stitch/montage area scans in Z-stack and/or time-lapse modes in your vessel of choice. For live cell imaging over multiple hours or days, the optional EVOS Onstage Incubator, which functions as an environmental chamber on top of the microscope stage, is operated seamlessly from within the instrument interface. The EVOS M7000 system is compatible with all accessories for the predecessor EVOS FL Auto imaging system.
Versatile and highly configurable
With a 5-position objective turret and the ability to simultaneous acquire images in four fluorescence channels plus transmitted light, the optical system can be easily configured to meet your needs. Choose from our broad range of high quality coverslip-corrected and long-working-distance objectives for use with plastic vessels or glass sides. Available from 1.25X to 100X magnification in achromat, fluorite, or apochromat models, together with over twenty LED light cubes, the sample imaging options are limitless. Dual monochrome and color high-sensitivity dual CMOS cameras enable seamless imaging of both fluorescent and chromogenically stained samples. Interchangeable vessel holders accommodate most vessel types and sizes, including slides, multi-well plates, culture flasks, and Petri dishes, and afford precise control and sample alignment by the automated stage. The vessel creation wizard in the user interface enables creation of profiles for custom sample vessels.
Powerful software
The EVOS M7000 software presents users with an extensive suite of tools for acquisition, visualization, and analysis. The user interface was designed with a strong emphasis on usability, including an option to view samples in a single field mode (Field View) or in a window that covers a much larger area (Area View). By using a zoom function, the Area View size can be adjusted. A user can explore a large sample area at low magnification, define regions of interest, and then seamlessly transition to the Scan function in Automate Mode and execute an area scan at higher magnification.
The enhanced power of the EVOS M7000 system enables more demanding automated acquisition routines like area scanning in time lapse and Z-stacks in scan mode to be performed. Visualization capabilities include multi-field image viewing gallery, mosaic tiling and tile stitching options, Z-stack visualization, and a video recording and movie maker/editor for time lapse sequences. The EVOS M7000 system is fully compatible with the EVOS Onstage Incubator environmental chamber for control of temperature, gases, and humidity. With the ability generate both hypoxic and normoxic conditions, it is possible to study live cells over extended periods with highly flexible time lapse automation routines.
For users needing more powerful image analysis capabilities, images acquired on the EVOS M7000 system can be analyzed directly using the optional Celleste Image Analysis Software. Celleste software offers powerful tools for image segmentation and classification that can be applied to a range of cell analysis applications for counting and measuring intensity, area, and shape changes over time, and more.
Smart LED illumination technology
All EVOS fluorescence imaging systems utilize our proprietary LED light cubes. This light engine outputs remarkable intensity over a short light path and delivers incomparable fluorophore excitation. Each light cube contains a precisely matched set of optical components to optimize the position, evenness, and intensity of the light beam. Digitally controlled LED light sources allow adjustment of illumination levels to the sample and experimental conditions to minimize the risk of photo-toxicity and photo-bleaching in time-lapse studies. Hard-coated filters give sharper edges and significantly higher transmission efficiencies than traditional soft-coated filters.
Specifications
Specifications
EVOS M7000 Imaging System
• 23 high‐resolution touchscreen Dell color monitor with 1920 x 1080 pixel resolution
• External Dell PC with an Intel Core i7-8700 processor, 32 GB DDR4 RAM, 512 GB PCIe SSD, and NVIDIA Quadro P1000 with 4 GB discrete video graphics running Windows 10, designed to operate with touchscreen monitor and microscope
EVOS M7000 Accessory Kit
• EVOS M7000 Quick Start Guide
• 16 GB USB 3.0 drive with user manual
• Universal power supply, 24V, 5A
• 3 North America (Type B) power cords*
• USB 3.0 type A to B cable
• Display port to display port cable, 6 ft
• Cube access tool
• EVOS calibration slide (AMEP4720)
• Vessel holder with retention for two slides (AMEPVH021)
• Vessel holder with retention for microwell plates (AMEPVH022). Also functions as adapter for vessel holders AMEPVH001 through AMEPVH018.
• Block condenser slider (AMEP4688)• Diffuser condenser slider for brightfield applications (AMEPDFS1)
• Slider for 4X objectives (AMEP4738)
• EVOS light shield
• Dust cover
• Shipping restraint removal guide
• Mouse pad
• Tech Support card
• Corded optical USB mouse
• Keyboard
* Note that three (3) country-specific power cord must be ordered separately in regions not using the Type B (North America) power plug:
AMEP4644 EVOS Power Cord, Type A (North America)
AMEP4645 EVOS Power Cord, Type G (United Kingdom)
AMEP4646 EVOS Power Cord, Type C (Europe)
AMEP4647 EVOS Power Cord, Type I (Australia)
AMEP4648 EVOS Power Cord, Type L (Italy)
AMEP4649 EVOS Power Cord, Type H (Israel)
AMEP4708 EVOS Power Cord, Type D (India/South Africa)
AMEP4782 Power Cord, Type I (China, CCC certified)
AMEP4786 Power Cord Type J (Switzerland)
EVOS
Room Temperature
1 Year Extended Warranty
EVOS™
System + SmartStart + 1 yr ext warranty
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.