Epredia™ Excelsior™ AS Tissue Processor Accessories
Maintain the Epredia™ Excelsior™ AS Tissue Processor with Epredia™ Excelsior AS Tissue Processor Accessories.
Supplier: Epredia™ A78410021
Description
For use with Shandon Excelsior ES Tissue ProcessorSpecifications
Random cassette basket | |
Excelsior AS Tissue Processor | |
Random basket processes up to 300 cassettes; not in organized fashion. |