Epredia™ Excelsior™ AS Tissue Processor Accessories
Maintain the Epredia™ Excelsior™ AS Tissue Processor with Epredia™ Excelsior AS Tissue Processor Accessories.
Supplier:  Epredia™ A82310038
 View more versions of this product
Catalog No. A82310038
Explore available promotions
Description
Description
Product Features:
  • Microwaveable design allowing pre-processing of tissue
  • Organized capacity of 300–cassettes in six basket-halves
Ordering Alerts:
Basket halves only match with each other but can be mixed with 222 cassette baskets or SecureSette Excelsior ES baskets
Includes:
Six 50-cassette basket halves and user guide and loading instructions
Compatible with:
Excelsior ES Tissue Processor
Specifications
Specifications
Organized Basket, 6 Sections
Excelsior AS Tissue Processor
300 Cassettes Kit
Ambient