Excelta™ 7000 Series Ultra Precision Wire Cutters with Medium Heads
For cutting leads and soft wire
Supplier:  Excelta™ 7242E
 View more versions of this product
Catalog No. 17456044
Explore available promotions
Description
Description
  • Induction hardened carbon tool steel
  • Edges hardened to RHC 63-65
  • Dual leaf springs
  • ESD safe cushion grips
Specifications
Specifications
Ergonomic short
Medium round
12.7 cm
0.003 to 0.032 in.
ESD-safe soft foam cushion
0.44 in.
1.11 cm
Ultra precision
5 in.
1% Carbon Tool Steel
Optimum flush
Set-screw, induction hardened cutting blades to RHC 63-65, Stainless Steel dual leaf springs
0.32 in.
0.81 cm