missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Excelta™ Diamond Tip Scribes

Aluminum handle

$157.59 - $157.59

Specifications

Product Type Diamond Tip Scribe
Tip Thickness 20 mm
Length (English) 4.5 in.
Length (Metric) 11.4 cm
View More Specs

Products 1
Catalog Number Mfr. No. Tip Style Price Quantity  
Catalog Number Mfr. No. Tip Style Price Quantity  
17467635
View Documents
Excelta™
475B
30° angled N/A
 
Description

Description

  • 20 mil tip size

Specifications

Specifications

Diamond Tip Scribe
20 mm
4.5 in.
11.4 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'