Learn More
Excelta™ General-Purpose Tweezers with Straight Tips with Medium Serrated Points
General-Purpose Tweezers with Straight Tips with Medium Serrated Points made of 400 series stainless steel provide durability.
Supplier:  Excelta™ 20ASSE
Catalog No. 17467231
Only null left
Explore available promotions
Description
Description
  • Inside and outside serration for better grip and handling small parts
  • Alignment pin for high strain applications maintains tip alignment
Specifications
Specifications
Straight, Medium Serrated
Stainless Steel
Tweezers, General Purpose
Alignment pin for high strain applications maintains tip alignment
0.03 cm
0.32 in.
Tweezers
4.5 in.
114 mm
1 Ea.
0.015 in.
General
0.81 cm
Product Suggestions
Product Suggestions