missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Excelta™ Precision Tweezers with Straight Strong Medium Tips

Precision Tweezers feature strong tips for long life.

Supplier:  Excelta™ 00CSA

 View more versions of this product

Catalog No. 18100802


Explore available promotions

Description

Description

Strong tips for long life

  • Anti-magnetic stainless steel or Carbon Steel
Specifications

Specifications

Strong Straight, Flame Hardened Blunt
4.25 in.
107 mm
1 Ea.
0.03 cm
0.32 in.
High-precision
00C
Anti-magnetic Stainless Steel
Tweezers, Electronic
0.015 in.
High
0.8 cm
SDS
Documents

Documents

Promotions

Promotions