missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

exo-7-Oxabicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0362006

 View more versions of this product

Catalog No. AAL0362006



Description

Description

Intermediate in a useful synthesis of 4,4-dialkyl-2-butenolides, based on reaction with two moles of Grignard reagent, followed by cycloreversion (elimination of furan) to release the double bond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Intermediate in a useful synthesis of 4,4-dialkyl-2-butenolides, based on reaction with two moles of Grignard reagent, followed by cycloreversion (elimination of furan) to release the double bond.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
Specifications

Specifications

exo-7-Oxabicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride
118°C to 119°C
MFCD00151506,MFCD00077442,MFCD00151506,MFCD00151506
10355
exo-3,6-epoxy-1,2,3,6-tetrahydrophthalic anhydride
QQYNRBAAQFZCLF-UHFFFAOYNA-N
4,10-dioxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
124671625
≥98%
6118-51-0
C8H6O4
5 g
Moisture sensitive
Hydrolyzes in water.
O=C1OC(=O)C2C3OC(C=C3)C12
166.13
166.13

Chemical Identifiers

6118-51-0
166.13
QQYNRBAAQFZCLF-UHFFFAOYNA-N
124671625
O=C1OC(=O)C2C3OC(C=C3)C12
C8H6O4
MFCD00151506,MFCD00077442,MFCD00151506,MFCD00151506
exo-3,6-epoxy-1,2,3,6-tetrahydrophthalic anhydride
4,10-dioxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 226-570-2

RTECSNumber : KC9650000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only