missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride, 98%, Thermo Scientific Chemicals

$136.79 - $432.81

Chemical Identifiers

CAS 29745-04-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00213361
InChI Key JAABVEXCGCXWRR-GUCUJZIJSA-N
Synonym exo-3,6-Epoxyhexahydrophthalic anhydride; Norcantharadin
PubChem CID 12251906
SMILES C1CC2C3C(C1O2)C(=O)OC3=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0294706
View Documents
Thermo Scientific Chemicals
L0294706
5 g
Each for $136.79
 
AAL0294714
View Documents
Thermo Scientific Chemicals
L0294714
25 g
Each for $432.81
 
Description

Description

exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride acts as a potential antitumor agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride acts as a potential antitumor agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Store under dry inert gas. Incompatible with Strong acids, Strong bases.
Specifications

Chemical Identifiers

29745-04-8
168.148
JAABVEXCGCXWRR-GUCUJZIJSA-N
12251906
C8H8O4
MFCD00213361
exo-3,6-Epoxyhexahydrophthalic anhydride; Norcantharadin
C1CC2C3C(C1O2)C(=O)OC3=O

Specifications

29745-04-8
C8H8O4
5 g
Moisture sensitive
Hydrolyzes in water.
C1CC2C3C(C1O2)C(=O)OC3=O
12251906
≥98%
111°C to 116°C
MFCD00213361
84287
exo-3,6-Epoxyhexahydrophthalic anhydride; Norcantharadin
JAABVEXCGCXWRR-GUCUJZIJSA-N
168.148
168.15
exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

RTECSNumber : RN8300000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only